Få mere at vide
N-Carbamoyl-2-methylbenzenesulfonamide (N-(Aminocarbonyl)-2-methylbenzenesulfonamide), Mikromol™
Artikelnummer. 30049318
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049318 25 mg
30050145 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049318 Leverandør Mikromol Leverandørnr. MM0325.110025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Carbamoyl-2-methylbenzenesulfonamide (N-(Aminocarbonyl)-2-methylbenzenesulfonamide)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Carbamoyl-2-methylbenzenesulfonamide
CAS 39051-77-9
Analyt- eller komponentnavne N-Carbamoyl-2-methylbenzenesulfonamide
Forsendelsestilstand Room Temperature
Molekylær formel C8 H10 N2 O3 S
Certificeringer/compliance ISO 17025
Synonym N-(Aminocarbonyl)-2-methylbenzenesulfonamide
InChI formel InChI=1S/C8H10N2O3S/c1-6-4-2-3-5-7(6)14(12,13)10-8(9)11/h2-5H,1H3,(H3,9,10,11)
SMIL Cc1ccccc1S(=O)(=O)NC(=O)N
IUPAC navn (2-methylphenyl)sulfonylurea
Molekylvægt (g/mol) 214.24
Mængde 25 mg
Formel vægt 214.0412 g/mol