Få mere at vide
N-Butyrylglycine-2,2-d2, CDN
Artikelnummer. 30321503
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321503 0.1 g 0.1g
30317343 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321503 Leverandør CDN Leverandørnr. CDND59850.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Butanoylglycine-D2 (glycine-2,2-D2)
CAS 1219799-10-6
Molekylær formel C6 D2 H9 N O3
Mængde 0.1 g
Synonym Glycine-2,2-d2, N-(1-oxobutyl)- (ACI), N-(1-Oxobutyl)glycine-2,2-d2 (ACI), N-Butyrylglycine-2,2-d2, N-Butanoylglycine-2,2-d2, N-Butanoylglycine-D2 (glycine-2,2-D2)
InChI formel InChI=1 S/C6H11NO3/c1-2-3-5(8)7-4-6(9)10/h2-4H2,1H3,(H,7,8)(H,9,10)/i4D2
SMIL [2 H]C([2 H])(NC(=O)CCC)C(=O)O
IUPAC navn 2-(butanoylamino)-2,2-dideuterioacetic acid
Molekylvægt (g/mol) 147.169
Formel vægt 147.086 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity