missing translation for 'onlineSavingsMsg'
Learn More

n-Butylmagnesium chloride, 1.5-2.5M in THF, Thermo Scientific™

Catalog Number p-7052213
Click to view available options
Quantity:
50 mL
250 mL
This item is not returnable. View return policy
693-04-9
C4H9ClMg
116.87
MFCD00000475
UUNGXYVLXYGZKH-UHFFFAOYSA-M
butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852
101837
magnesium;butane;chloride
CCCC[Mg]Cl
This item is not returnable. View return policy
693-04-9
C4H9ClMg
116.87
MFCD00000475
UUNGXYVLXYGZKH-UHFFFAOYSA-M
butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852
101837
magnesium;butane;chloride
CCCC[Mg]Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Brown
Odor Ether-like
Linear Formula CH3(CH2)3MgCl
Quantity 50 mL
UN Number UN3399
Beilstein 3587228
Sensitivity Air and moisture sensitive
Formula Weight 116.87
Concentration or Composition (by Analyte or Components) 1.5 to 2.5M in THF
Physical Form Liquid
Chemical Name or Material n-Butylmagnesium chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'