Få mere at vide
N-Butyldeoxynojirimycin Hydrochloride-d9, TRC
Artikelnummer. 30345007
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30345007 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345007 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69100210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Butyldeoxynojirimycin Hydrochloride-d9
CAS 1883545-57-0
Molekylær formel C10H13D9ClNO4
Mængde 10 mg
Synonym (2R,3R,4R,5S)-1-Butyl-2-(hydroxymethyl)-,3,4,5-Piperidinetriol Hydrochloride-d9, Miglustat Hydrochloride-d9
InChI formel InChI=1S/C10H21NO4.ClH/c1-2-3-4-11-5-8(13)10(15)9(14)7(11)6-12;/h7-10,12-15H,2-6H2,1H3;1H/t7-,8+,9-,10-;/m1./s1/i1D3,2D2,3D2,4D2;
SMIL O[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)N(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C1.Cl
IUPAC navn (2R,3R,4R,5S)-2-(hydroxymethyl)-1-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)piperidine-3,4,5-triol;hydrochloride
Molekylvægt (g/mol) 264.79