Få mere at vide
n-Butyl-d9-amine HCl, TRC
Artikelnummer. 30334582
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30334582 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334582 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6931215MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Butyl-d9-amine HCl
CAS 344298-86-8
Molekylær formel C4 D9 H2 N . Cl H
Mængde 5 mg
Synonym 1-Butan-1,1,2,2,3,3,4,4,4-d9-amine, hydrochloride (9CI), 1-Butan-1,1,2,2,3,3,4,4,4-d9-amine hydrochloride, 1-Butylamine-D9 hydrochloride, n-Butyl-d9-amine hydrochloride, 1-Butanamine-d9 hydrochloride
InChI formel InChI=1S/C4H11N.ClH/c1-2-3-4-5;/h2-5H2,1H3;1H/i1D3,2D2,3D2,4D2;
SMIL Cl.[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])N
IUPAC navn 1,1,2,2,3,3,4,4,4-nonadeuteriobutan-1-amine;hydrochloride
Molekylvægt (g/mol) 118.653
Formel vægt 118.122