Få mere at vide
n-Butyl Alcohol-OD, CDN
Artikelnummer. 30326851
Skift visning
Mængde:
25 g
Artikelnummer. Mængde
30326851 25 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326851 Leverandør CDN Leverandørnr. CDND020225G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Butanol-d
CAS 4712-38-3
Molekylær formel C4 D H9 O
Mængde 25 g
Synonym 1-Butanol-d (9 CI), Butyl alcohol-d (6 CI, 7 CI, 8 CI), 1-Deuteriooxybutane, Butanol-O-d, n-Butanol-O-d, 1-Butanol-O-D, n-Butyl alcohol-OD
InChI formel InChI=1 S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3/i5D
SMIL [2 H]OCCCC
IUPAC navn 1-deuteriooxybutane
Molekylvægt (g/mol) 75.128
Formel vægt 75.079 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity