missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4228476
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
123-86-4
C6H12O2
116.16
MFCD00009445
DKPFZGUDAPQIHT-UHFFFAOYSA-N
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
31272
CHEBI:31328
butyl acetate
CCCCOC(C)=O
This item is not returnable. View return policy
123-86-4
C6H12O2
116.16
MFCD00009445
DKPFZGUDAPQIHT-UHFFFAOYSA-N
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
31272
CHEBI:31328
butyl acetate
CCCCOC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -78.0°C
Density 0.8800g/mL
Boiling Point 127.0°C
Flash Point 27°C
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3CO2(CH2)3CH3
Refractive Index 1.3930 to 1.3950
Quantity 100 mL
Merck Index 15,1537
Specific Gravity 0.88
Solubility Information Solubility in water: 7g/L (20°C)
Viscosity 0.69 mPa.s (25°C)
Formula Weight 116.16
Percent Purity 99+%
Grade Extra Dry
Chemical Name or Material n-Butyl acetate, Extra Dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'