Få mere at vide
n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4, TRC
Artikelnummer. 30334814
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30334814 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334814 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6936041MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4
CAS 1219798-67-0
Molekylær formel C11 D4 H10 O3
Mængde 1 mg
Synonym Benzoic-2,3,5,6-d4 acid, 4-hydroxy-, butyl ester (ACI), Butyl 4-hydroxybenzoate-2,3,5,6-d4 (ACI), 4-Hydroxybenzoic-2,3,5,6-d4 acid butyl ester, n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4, Butyl Parahydroxybenzoate-d4, Butylparaben-d4, Butyl Parahydroxybenzoate-D4 (4-hydroxybenzoic-2,3,5,6-D4)
InChI formel InChI=1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3/i4D,5D,6D,7D
SMIL [2H]c1c([2H])c(C(=O)OCCCC)c([2H])c([2H])c1O
IUPAC navn butyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate
Molekylvægt (g/mol) 198.252
Formel vægt 198.119