Få mere at vide
n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4, CDN
Artikelnummer. 30314444
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314444 0.05 g -
30326076 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314444 Leverandør CDN Leverandørnr. CDND72350.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4
CAS 1219798-67-0
Molekylær formel C11 D4 H10 O3
Mængde 0.05 g
Synonym Benzoic-2,3,5,6-d4 acid, 4-hydroxy-, butyl ester (ACI), Butyl 4-hydroxybenzoate-2,3,5,6-d4 (ACI), 4-Hydroxybenzoic-2,3,5,6-d4 acid butyl ester, n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4, Butyl Parahydroxybenzoate-d4, Butylparaben-d4, Butyl Parahydroxybenzoate-D4 (4-hydroxybenzoic-2,3,5,6-D4)
InChI formel InChI=1 S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12 H,2-3,8H2,1H3/i4D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c(C(=O)OCCCC)c([2 H])c([2 H])c1O
IUPAC navn butyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate
Molekylvægt (g/mol) 198.252
Formel vægt 198.119 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity