Learn More
Alfa Aesar™ N-Boc-sarcosine methyl ester, 97%
Brand:  Alfa Aesar™ H62304.MD
Additional Details : CAS Number : 42492-57-9 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15488838
  • 243.13 DKK / 250mg
Estimated Shipment
26-10-2020
Add to basket
Description
Description
Applications
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.
Notes
Incompatible with oxidizing agents. Heat sensitive.
Specifications
Chemical Identifiers
42492-57-9
203.238
LTBDTYJYKRLTRN-UHFFFAOYSA-N
14018930
CC(C)(C)OC(=O)N(C)CC(=O)OC
C9H17NO4
MFCD04973115
methyl 2-tert-butoxycarbonyl methyl amino acetate, n-boc-sarcosine methyl ester, boc-gly-ome, boc-sarcosine methyl ester, ltbdtyjykrltrn-uhfffaoysa, n-t-butoxycarbonyl-sarcosine methyl ester, n-tert-butoxycarbonylsarcosine methyl ester, methyl n-tert-butoxycarbonyl-n-methylglycinate, methyl 2-tert-butoxy carbonyl methyl amino acetate
methyl 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetate
Specifications
N-Boc-sarcosine methyl ester
C9H17NO4
MFCD04973115
LTBDTYJYKRLTRN-UHFFFAOYSA-N
methyl 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetate
14018930
Liquid
Yellow
42492-57-9
(CH3)3COCON(CH3)CH2CO2CH3
methyl 2-tert-butoxycarbonyl methyl amino acetate, n-boc-sarcosine methyl ester, boc-gly-ome, boc-sarcosine methyl ester, ltbdtyjykrltrn-uhfffaoysa, n-t-butoxycarbonyl-sarcosine methyl ester, n-tert-butoxycarbonylsarcosine methyl ester, methyl n-tert-butoxycarbonyl-n-methylglycinate, methyl 2-tert-butoxy carbonyl methyl amino acetate
CC(C)(C)OC(=O)N(C)CC(=O)OC
203.238
203.24
97%
250mg
Videos
SDS Documents