Få mere at vide
N-Boc-N-methylpiperazine-d3, TRC
Artikelnummer. 30338713
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30338713 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338713 Leverandør Toronto Research Chemicals Leverandørnr. TRCB65697325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Boc-N-methylpiperazine-d3
CAS 1459230-58-0
Molekylær formel C10H17D3N2O2
Mængde 25 mg
Synonym 4-Methyl-1-piperazinecarboxylic Acid 1,1-Dimethylethyl Ester-d3, 1-Methyl-4-(tert-butoxycarbonyl)piperazine, 4-Methylpiperazine-1-carboxylic Acid tert-Butyl Ester-d3
InChI formel InChI=1S/C10H20N2O2/c1-10(2,3)14-9(13)12-7-5-11(4)6-8-12/h5-8H2,1-4H3/i4D3
SMIL [2H]C([2H])([2H])N1CCN(CC1)C(=O)OC(C)(C)C
IUPAC navn tert-butyl 4-(trideuteriomethyl)piperazine-1-carboxylate
Molekylvægt (g/mol) 203.3
Formel vægt 203.1713