missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-N-methyl-DL-valine, 98%

Brand:  Alfa Aesar™ H62591.03

Additional Details : CAS Number : 13850-91-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469098

  • 1498.71 DKK / 1g


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Specifications

Chemical Identifiers

13850-91-4
231.292
XPUAXAVJMJDPDH-UHFFFAOYSA-N
4003029
CC(C)C(C(=O)O)N(C)C(=O)OC(C)(C)C
C11H21NO4
MFCD00153317
boc-n-me-dl-val-oh, valine,n-1,1-dimethylethoxy carbonyl-n-methyl, dl-valine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, 3-methyl-2-methyl-2-methylpropan-2-yl oxycarbonyl amino butanoic acid, n-tert-butoxycarbonyl-n-methylvaline, n-methyl-n-2-methyl-2-propanyl oxy carbonyl valine, 2-tert-butoxy carbonyl methyl amino-3-methylbutanoic acid
3-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]butanoic acid

Specifications

N-Boc-N-methyl-DL-valine
C11H21NO4
boc-n-me-dl-val-oh, valine,n-1,1-dimethylethoxy carbonyl-n-methyl, dl-valine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, 3-methyl-2-methyl-2-methylpropan-2-yl oxycarbonyl amino butanoic acid, n-tert-butoxycarbonyl-n-methylvaline, n-methyl-n-2-methyl-2-propanyl oxy carbonyl valine, 2-tert-butoxy carbonyl methyl amino-3-methylbutanoic acid
CC(C)C(C(=O)O)N(C)C(=O)OC(C)(C)C
231.292
231.29
98%
13850-91-4
MFCD00153317
XPUAXAVJMJDPDH-UHFFFAOYSA-N
3-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]butanoic acid
4003029
Solid
1g
Videos
SDS