Få mere at vide
N-Boc-N-deshydroxyethyl Dasatinib-d8, TRC
Artikelnummer. 30331628
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30331628 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331628 Leverandør Toronto Research Chemicals Leverandørnr. TRCB64630710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Boc-N-deshydroxyethyl Dasatinib-d8
CAS 1263379-04-9
Molekylær formel C25H22D8ClN7O3S
Mængde 10 mg
InChI formel InChI=1S/C25H30ClN7O3S/c1-15-7-6-8-17(26)21(15)31-22(34)18-14-27-23(37-18)30-19-13-20(29-16(2)28-19)32-9-11-33(12-10-32)24(35)36-25(3,4)5/h6-8,13-14H,9-12H2,1-5H3,(H,31,34)(H,27,28,29,30)/i9D2,10D2,11D2,12D2
SMIL [2H]C1([2H])N(C(=O)OC(C)(C)C)C([2H])([2H])C([2H])([2H])N(c2cc(Nc3ncc(s3)C(=O)Nc4c(C)cccc4Cl)nc(C)n2)C1([2H])[2H]
IUPAC navn tert-butyl 4-[6-[[5-[(2-chloro-6-methylphenyl)carbamoyl]-1,3-thiazol-2-yl]amino]-2-methylpyrimidin-4-yl]-2,2,3,3,5,5,6,6-octadeuteriopiperazine-1-carboxylate
Molekylvægt (g/mol) 552.12