missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Boc-N-desethyl Acetildenafil, TRC

Artikelnummer. 30330563
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30330563 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30330563 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6462805MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-Boc-N-desethyl Acetildenafil
CAS 1246820-46-1
Molekylær formel C28 H38 N6 O5
Mængde 5 mg
Synonym 4-[2-[3-(6,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]-2-oxoethyl]-1-piperazinecarboxylic acid 1,1-dimethylethyl ester, N-Boc-N-desethyl Acetildenafil
InChI formel InChI=1S/C28H38N6O5/c1-7-9-20-23-24(32(6)31-20)26(36)30-25(29-23)19-16-18(10-11-22(19)38-8-2)21(35)17-33-12-14-34(15-13-33)27(37)39-28(3,4)5/h10-11,16H,7-9,12-15,17H2,1-6H3,(H,29,30,36)
SMIL CCCc1nn(C)c2C(=O)N=C(Nc12)c3cc(ccc3OCC)C(=O)CN4CCN(CC4)C(=O)OC(C)(C)C
IUPAC navn tert-butyl 4-[2-[4-ethoxy-3-(1-methyl-7-oxo-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-5-yl)phenyl]-2-oxoethyl]piperazine-1-carboxylate
Molekylvægt (g/mol) 538.64
Formel vægt 538.2904
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.