missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
N-Boc-N-desethyl Acetildenafil, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Boc-N-desethyl Acetildenafil |
| CAS | 1246820-46-1 |
| Molekylær formel | C28 H38 N6 O5 |
| Mængde | 50 mg |
| Synonym | 4-[2-[3-(6,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]-2-oxoethyl]-1-piperazinecarboxylic acid 1,1-dimethylethyl ester, N-Boc-N-desethyl Acetildenafil |
| InChI formel | InChI=1S/C28H38N6O5/c1-7-9-20-23-24(32(6)31-20)26(36)30-25(29-23)19-16-18(10-11-22(19)38-8-2)21(35)17-33-12-14-34(15-13-33)27(37)39-28(3,4)5/h10-11,16H,7-9,12-15,17H2,1-6H3,(H,29,30,36) |
| SMIL | CCCc1nn(C)c2C(=O)N=C(Nc12)c3cc(ccc3OCC)C(=O)CN4CCN(CC4)C(=O)OC(C)(C)C |
| IUPAC navn | tert-butyl 4-[2-[4-ethoxy-3-(1-methyl-7-oxo-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-5-yl)phenyl]-2-oxoethyl]piperazine-1-carboxylate |
| Molekylvægt (g/mol) | 538.64 |
| Formel vægt | 538.2904 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?