Få mere at vide
N’-Boc-N-Benzylpiperazine-d8, TRC
Artikelnummer. 30331543
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30331543 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331543 Leverandør Toronto Research Chemicals Leverandørnr. TRCB626302100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N'-Boc-N-Benzylpiperazine-d8
CAS 1216387-74-4
Molekylær formel C16 H16 D8 N2 O2
Mængde 100 mg
Synonym 4-(Phenylmethyl-d8)-1-piperazinecarboxylic Acid 1,1-Dimethylethyl Ester, 1-(tert-Butoxycarbonyl)-4-(phenylmethyl)piperazine-d8, 4-Benzyl-1-piperazinecarboxylate-d8
InChI formel InChI=1S/C16H24N2O2/c1-16(2,3)20-15(19)18-11-9-17(10-12-18)13-14-7-5-4-6-8-14/h4-8H,9-13H2,1-3H3/i9D2,10D2,11D2,12D2
SMIL [2H]C1([2H])N(Cc2ccccc2)C([2H])([2H])C([2H])([2H])N(C(=O)OC(C)(C)C)C1([2H])[2H]
IUPAC navn tert-butyl 4-benzyl-2,2,3,3,5,5,6,6-octadeuteriopiperazine-1-carboxylate
Molekylvægt (g/mol) 284.42
Formel vægt 284.234