missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-N,2-Dimethylalanine, 98+%, Thermo Scientific Chemicals

Catalog Number p-2691903
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
146000-39-7
C10H19NO4
217.27
MFCD01862912
KPDLITGXUYMJEC-UHFFFAOYSA-N
boc-n-me-aib-oh, boc-n,2-dimethylalanine, boc-n-methyl-aminoisobutyric acid, 2-tert-butoxycarbonyl methyl amino-2-methylpropanoic acid, boc-n-methyl-aib, acmc-1c0qg, n-boc-n,2-dimethylalanine, n-boc-n,2-dimethyl-dl-alanine, 2-methyl-2-methyl-2-methylpropan-2-yl oxycarbonyl amino propanoic acid, n-boc-, a-methylamino isobutyric acid
2734752
2-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
CN(C(=O)OC(C)(C)C)C(C)(C)C(O)=O
This item is not returnable. View return policy
146000-39-7
C10H19NO4
217.27
MFCD01862912
KPDLITGXUYMJEC-UHFFFAOYSA-N
boc-n-me-aib-oh, boc-n,2-dimethylalanine, boc-n-methyl-aminoisobutyric acid, 2-tert-butoxycarbonyl methyl amino-2-methylpropanoic acid, boc-n-methyl-aib, acmc-1c0qg, n-boc-n,2-dimethylalanine, n-boc-n,2-dimethyl-dl-alanine, 2-methyl-2-methyl-2-methylpropan-2-yl oxycarbonyl amino propanoic acid, n-boc-, a-methylamino isobutyric acid
2734752
2-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
CN(C(=O)OC(C)(C)C)C(C)(C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 153°C to 157°C
CAS Min % 98.0
CAS Max % 100.0
Color White
Assay Percent Range 98+%
Infrared Spectrum Authentic
Linear Formula (CH3)3COCON(CH3)C(CH3)2COOH
Packaging Glass Bottle
Formula Weight 217.26
Percent Purity ≥98%
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-N, 2-Dimethylalanine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'