missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-phenylalanyl-L-phenylalanine, 95%

Brand:  Alfa Aesar™ H63788.03

Additional Details : CAS Number : 13122-90-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15490669

  • 429.58 DKK / 1g


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

13122-90-2
412.486
NNOBHAOOLCEJBL-OALUTQOASA-N
7019052
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC2=CC=CC=C2)C(=O)O
C23H28N2O5
MFCD00190824
boc-phe-phe-oh, boc-l-diphenylalanine, s-2-s-2-tert-butoxycarbonyl amino-3-phenylpropanamido-3-phenylpropanoic acid, n-tert-butoxycarbonyl-l-phenylalanyl-l-phenylalanine, 2s-2-2s-2-tert-butoxycarbonyl amino-3-phenylpropanamido-3-phenylpropanoic acid, n-boc-l-phenylalanyl-l-phenylalanine, n-n-tert-butoxycarbonyl-l-phenylalanyl-l-phenylalanine
(2S)-2-[[(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoyl]amino]-3-phenylpropanoic acid

Specifications

N-Boc-L-phenylalanyl-L-phenylalanine
C23H28N2O5
boc-phe-phe-oh, boc-l-diphenylalanine, s-2-s-2-tert-butoxycarbonyl amino-3-phenylpropanamido-3-phenylpropanoic acid, n-tert-butoxycarbonyl-l-phenylalanyl-l-phenylalanine, 2s-2-2s-2-tert-butoxycarbonyl amino-3-phenylpropanamido-3-phenylpropanoic acid, n-boc-l-phenylalanyl-l-phenylalanine, n-n-tert-butoxycarbonyl-l-phenylalanyl-l-phenylalanine
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC2=CC=CC=C2)C(=O)O
412.486
412.48
95%
13122-90-2
MFCD00190824
NNOBHAOOLCEJBL-OALUTQOASA-N
(2S)-2-[[(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoyl]amino]-3-phenylpropanoic acid
7019052
Solid
1g
Videos
SDS Documents