Få mere at vide
N-Boc-L-glutamic Acid Alpha-Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Boc-L-glutamic Acid alpha-Methyl Ester |
| CAS | 72086-72-7 |
| Molekylær formel | C11 H19 N O6 |
| Mængde | 5 g |
| Synonym | N-Boc-L-glutamic Acid α-Methyl Ester, N-[(1,1-Dimethylethoxy)carbonyl]-L-glutamic Acid 1-Methyl Ester, N-(tert-Butoxycarbonyl)glutamic Acid α-Methyl Ester |
| InChI formel | InChI=1S/C11H19NO6/c1-11(2,3)18-10(16)12-7(9(15)17-4)5-6-8(13)14/h7H,5-6H2,1-4H3,(H,12,16)(H,13,14)/t7-/m0/s1 |
| SMIL | COC(=O)[C@H](CCC(=O)O)NC(=O)OC(C)(C)C |
| IUPAC navn | (4S)-5-methoxy-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
| Molekylvægt (g/mol) | 261.27 |
| Formel vægt | 261.1212 |