Learn More
Alfa Aesar™ N-Boc-L-glutamic acid 5-(9-fluorenylmethyl) ester, 98%
Brand:  Alfa Aesar™ H63101.03
Additional Details : CAS Number : 123417-18-5 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15408126
  • 751.66 DKK / 1g
Estimated Shipment date
from Supplier 01-08-2018
Add to basket
Specifications
Chemical Identifiers
N-Boc-L-glutamic acid 5-(9-fluorenylmethyl) ester
425.47
Boc-Glu(Ofm)-OH
123417-18-5
MFCD00076934
Specifications
N-Boc-L-glutamic acid 5-(9-fluorenylmethyl) ester
Solid
425.47
Boc-Glu(Ofm)-OH
123417-18-5
98%
MFCD00076934
1g