missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycyl-L-leucine, 98%, Thermo Scientific™

Catalog Number 15489078
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
51871-42-2
C13H24N2O5
288.34
MFCD00190801
RGNNFKYULPHSJI-IMWMWJONNA-N
boc-gly-leu-oh, l-leucine, n-1,1-dimethylethoxy carbonyl glycyl, boc-gly-l-leu-oh, 2s-2-2-tert-butoxycarbonyl amino acetamido-4-methylpentanoic acid
11953699
(2S)-4-methyl-2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]pentanoic acid
CC(C)C[C@H](NC(=O)CNC(=O)OC(C)(C)C)C(O)=O
This item is not returnable. View return policy
51871-42-2
C13H24N2O5
288.34
MFCD00190801
RGNNFKYULPHSJI-IMWMWJONNA-N
boc-gly-leu-oh, l-leucine, n-1,1-dimethylethoxy carbonyl glycyl, boc-gly-l-leu-oh, 2s-2-2-tert-butoxycarbonyl amino acetamido-4-methylpentanoic acid
11953699
(2S)-4-methyl-2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]pentanoic acid
CC(C)C[C@H](NC(=O)CNC(=O)OC(C)(C)C)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 288.34
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Boc-glycyl-L-leucine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'