Få mere at vide
N-Boc-erythro-sphingosine, TRC
Artikelnummer. 30179241
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179241 25 mg
30179066 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179241 Leverandør Toronto Research Chemicals Leverandørnr. TRCB66775025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 116467-63-1
Molekylær formel C23 H45 N O4
Molekylvægt (g/mol) 399.61
IUPAC navn tert-butyl N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]carbamate
SMIL CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(=O)OC(C)(C)C
Tekniske data
Kemisk navn eller materiale N-Boc-erythro-sphingosine
InChI formel InChI=1S/C23H45NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-21(26)20(19-25)24-22(27)28-23(2,3)4/h17-18,20-21,25-26H,5-16,19H2,1-4H3,(H,24,27)/b18-17+/t20-,21+/m0/s1
Mængde 25 mg
Formel vægt 399.3349