missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-DL-alanine, 97%, Thermo Scientific Chemicals

Catalog Number p-7041480
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
3744-87-4
C8H15NO4
189.211
MFCD00063124
QVHJQCGUWFKTSE-UHFFFAOYSA-N
boc-dl-alanine, boc-dl-ala-oh, 2-tert-butoxycarbonyl amino propanoic acid, n-boc-dl-alanine, 2-n-boc-amino-propionic acid, n-tert-butoxycarbonyl alanine, 2-tert-butoxycarbonylamino-propionic acid, 2-tert-butoxycarbonylamino propanoic acid, 2-tert-butoxy carbonyl amino propanoic acid
268471
2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C(=O)O)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
3744-87-4
C8H15NO4
189.211
MFCD00063124
QVHJQCGUWFKTSE-UHFFFAOYSA-N
boc-dl-alanine, boc-dl-ala-oh, 2-tert-butoxycarbonyl amino propanoic acid, n-boc-dl-alanine, 2-n-boc-amino-propionic acid, n-tert-butoxycarbonyl alanine, 2-tert-butoxycarbonylamino-propionic acid, 2-tert-butoxycarbonylamino propanoic acid, 2-tert-butoxy carbonyl amino propanoic acid
268471
2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C(=O)O)NC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material N-Boc-DL-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'