missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-D-glutamic acid 1-tert-butyl ester, 95%

Brand:  Alfa Aesar™ H62320.03

Additional Details : CAS Number : 73872-71-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478848

  • 1491.59 DKK / 1g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

73872-71-6
303.355
YMOYURYWGUWMFM-SECBINFHSA-N
7018746
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OC(C)(C)C
C14H25NO6
MFCD00076926
boc-d-glu-otbu, r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, 4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-o-t-bu, n-boc-d-glutamic acid 1-tert-butyl ester, d-glutamicacid,n-1,1-dimethylethoxy carbonyl-,1-1,1-dimethylethyl ester
(4R)-5-[(2-methylpropan-2-yl)oxy]-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid

Specifications

N-Boc-D-glutamic acid 1-tert-butyl ester
C14H25NO6
boc-d-glu-otbu, r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, 4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-o-t-bu, n-boc-d-glutamic acid 1-tert-butyl ester, d-glutamicacid,n-1,1-dimethylethoxy carbonyl-,1-1,1-dimethylethyl ester
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OC(C)(C)C
303.355
303.35
95%
1g
73872-71-6
MFCD00076926
YMOYURYWGUWMFM-SECBINFHSA-N
(4R)-5-[(2-methylpropan-2-yl)oxy]-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
7018746
Powder
Brown to White
Videos
SDS Documents