missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
N-Boc-4-methylene-L-proline, 97%, Thermo Scientific™
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
As important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry. The fluorous prolins were synthesized from commercially available N-Boc-4-methylene- L-proline.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
As important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry. The fluorous prolins were synthesized from commercially available N-Boc-4-methylene- L-proline.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Boc-4-methylene-L-proline |
| Smeltepunkt | 110°C to 114°C |
| Mængde | 250 mg |
| Opløselighedsinformation | Insoluble in water. |
| Formel vægt | 227.26 |
| Procent renhed | 97% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?