missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-chloro-L-phenylalanine, 95%, Thermo Scientific™

Catalog Number p-7041195
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
68090-88-0
C14H17ClNO4
298.74
MFCD00057808
BETBOAZCLSJOBQ-NSHDSACASA-M
boc-phe 4-cl-oh, boc-l-4-chlorophenylalanine, boc-4-chloro-l-phenylalanine, boc-l-4-chlorophe, boc-l-phe 4-cl-oh, n-boc-4-chloro-l-phenylalanine, n-tert-butoxycarbonyl-4-chloro-l-phenylalanine, s-n-boc-4-chlorophenylalanine, s-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, boc-p-chloro-phe-oh
2734499
(2S)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C=C1)C([O-])=O
This item is not returnable. View return policy
68090-88-0
C14H17ClNO4
298.74
MFCD00057808
BETBOAZCLSJOBQ-NSHDSACASA-M
boc-phe 4-cl-oh, boc-l-4-chlorophenylalanine, boc-4-chloro-l-phenylalanine, boc-l-4-chlorophe, boc-l-phe 4-cl-oh, n-boc-4-chloro-l-phenylalanine, n-tert-butoxycarbonyl-4-chloro-l-phenylalanine, s-n-boc-4-chlorophenylalanine, s-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, boc-p-chloro-phe-oh
2734499
(2S)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C=C1)C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼110°C
Quantity 1 g
Beilstein 4261946
Formula Weight 299.75
Percent Purity 95%
Chemical Name or Material N-Boc-4-chloro-L-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'