Få mere at vide
N-Boc-4-[bis(2-hydroxyethyl-d4)amino]-L-phenylalanine Methyl Ester, TRC
Artikelnummer. 30345927
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30345927 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345927 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6432021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Boc-4-[bis(2-hydroxyethyl-d4)amino]-L-phenylalanine Methyl Ester
CAS 1217602-23-7
Molekylær formel C19H22D8N2O6
Mængde 1 mg
Synonym N-(tert-Butoxycarbonyl)-4-[bis(2-hydroxyethyl-d4)amino]-L-phenylalanine Methyl Ester
InChI formel InChI=1S/C19H30N2O6/c1-19(2,3)27-18(25)20-16(17(24)26-4)13-14-5-7-15(8-6-14)21(9-11-22)10-12-23/h5-8,16,22-23H,9-13H2,1-4H3,(H,20,25)/t16-/m0/s1/i9D2,10D2,11D2,12D2
SMIL [2H]C([2H])(O)C([2H])([2H])N(c1ccc(C[C@H](NC(=O)OC(C)(C)C)C(=O)OC)cc1)C([2H])([2H])C([2H])([2H])O
IUPAC navn methyl (2S)-3-[4-[bis(1,1,2,2-tetradeuterio-2-hydroxyethyl)amino]phenyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
Molekylvægt (g/mol) 390.5
Formel vægt 390.2606