missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-hydroxy-L-valine, 97%, Thermo Scientific Chemicals

Catalog Number p-7034731
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
102507-13-1
C10H19NO5
233.26
MFCD03094792
SZVRVSZFEDIMFM-ZCFIWIBFSA-N
s-2-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-n-boc-amino-3-hydroxy-3-methylbutyric acid, s-n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, n-tert-butoxycarbonyl-3-hydroxy-l-valine, 2s-2-n-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-boc-amino-3-hydroxy-3-methylbutyric acid, boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-3-hydroxy
11075314
(2S)-3-hydroxy-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)O
This item is not returnable. View return policy
102507-13-1
C10H19NO5
233.26
MFCD03094792
SZVRVSZFEDIMFM-ZCFIWIBFSA-N
s-2-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-n-boc-amino-3-hydroxy-3-methylbutyric acid, s-n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, n-tert-butoxycarbonyl-3-hydroxy-l-valine, 2s-2-n-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-boc-amino-3-hydroxy-3-methylbutyric acid, boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-3-hydroxy
11075314
(2S)-3-hydroxy-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 233.26
Percent Purity 97%
Chemical Name or Material N-Boc-3-hydroxy-L-valine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'