missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-cyclopropyl-L-glycine, 95%, Thermo Scientific Chemicals

Catalog Number p-7054587
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
155976-13-9
C10H17NO4
215.25
MFCD06659132,MFCD01861823
QFVJNEASAAJIDF-ZETCQYMHSA-N
boc-l-cyclopropylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopropylacetic acid, boc-s-cyclopropylglycine, s-tert-butoxycarbonylamino-cyclopropyl-acetic acid, s-boc-amino-cyclopropylacetic acid, 2s-2-amino-2-cyclopropylethanoic acid, n-boc protected, s-2-tert-butoxycarbonylamino-2-cyclopropylacetic acid, s-tert-butoxycarbonyl amino cyclopropyl acetic acid, boc-l-cyclopropyl glycine
2761461
(2S)-2-cyclopropyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O
This item is not returnable. View return policy
155976-13-9
C10H17NO4
215.25
MFCD06659132,MFCD01861823
QFVJNEASAAJIDF-ZETCQYMHSA-N
boc-l-cyclopropylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopropylacetic acid, boc-s-cyclopropylglycine, s-tert-butoxycarbonylamino-cyclopropyl-acetic acid, s-boc-amino-cyclopropylacetic acid, 2s-2-amino-2-cyclopropylethanoic acid, n-boc protected, s-2-tert-butoxycarbonylamino-2-cyclopropylacetic acid, s-tert-butoxycarbonyl amino cyclopropyl acetic acid, boc-l-cyclopropyl glycine
2761461
(2S)-2-cyclopropyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Beilstein 6650020
Formula Weight 215.25
Percent Purity 95%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material N-Boc-2-cyclopropyl-L-glycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'