missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-2,4-dichloro-D-phenylalanine, 98%

Brand:  Alfa Aesar™ H62706.03

Additional Details : CAS Number : 114873-12-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469188

  • 1297.39 DKK / 1g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

114873-12-0
334.193
KSDWBXFRQWMWHO-LLVKDONJSA-N
7010031
CC(C)(C)OC(=O)NC(CC1=C(C=C(C=C1)Cl)Cl)C(=O)O
C14H17Cl2NO4
MFCD01862941
r-2-tert-butoxycarbonyl amino-3-2,4-dichlorophenyl propanoic acid, boc-d-2,4-dichlorophenylalanine, boc-2,4-dichloro-d-phenylalanin, boc-d-2,4-dichlorophe, boc-2,4-dichloro-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-2,4-dichlorophenyl propanoic acid, boc-d-phe 2,4-dicl-oh, d-phenylalanine, 2,4-dichloro-n-1,1-dimethylethoxy carbonyl, 2r-3-2,4-dichlorophenyl-2-tert-butoxy carbonylamino propanoic acid, r-2-tert-butoxycarbonylamino-3-2,4-dichloro-phenyl-propionic acid
(2R)-3-(2,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

N-Boc-2,4-dichloro-D-phenylalanine
C14H17Cl2NO4
r-2-tert-butoxycarbonyl amino-3-2,4-dichlorophenyl propanoic acid, boc-d-2,4-dichlorophenylalanine, boc-2,4-dichloro-d-phenylalanin, boc-d-2,4-dichlorophe, boc-2,4-dichloro-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-2,4-dichlorophenyl propanoic acid, boc-d-phe 2,4-dicl-oh, d-phenylalanine, 2,4-dichloro-n-1,1-dimethylethoxy carbonyl, 2r-3-2,4-dichlorophenyl-2-tert-butoxy carbonylamino propanoic acid, r-2-tert-butoxycarbonylamino-3-2,4-dichloro-phenyl-propionic acid
CC(C)(C)OC(=O)NC(CC1=C(C=C(C=C1)Cl)Cl)C(=O)O
334.193
334.2
98%
1g
114873-12-0
MFCD01862941
KSDWBXFRQWMWHO-LLVKDONJSA-N
(2R)-3-(2,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
7010031
Solid
133°C to 137°C
Videos
SDS