Få mere at vide
N-Boc-2-(2-naphthyl)-D-alanine, 99%, Thermo Scientific™
Artikelnummer. 15478427
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15478427 1 g 1g
15488427 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15478427 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H51968.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76985-10-9
Molekylær formel C18H21NO4
Molekylvægt (g/mol) 315.37
MDL nummer MFCD00076900
InChI nøgle URKWHOVNPHQQTM-OAHLLOKOSA-N
Synonym boc-d-2-nal-oh, boc-3-2-naphthyl-d-alanine, n-tert-butoxycarbonyl-3-2-naphthyl-d-alanine, boc-2-d-nal-oh, r-n-boc-2-naphthylalanine, boc-d-3-2-naphthyl-alanine, n-boc-3-2-naphthyl-d-alanine, n-boc-2-2-naphthyl-d-alanine, r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid
PubChem CID 2734481
IUPAC navn (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-2-ylpropanoic acid
SMIL CC(C)(C)OC(=O)N[C@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O
Tekniske data
Kemisk navn eller materiale N-Boc-2-(2-naphthyl)-D-alanine
Smeltepunkt 94°C to 96°C
Beilstein 4200660
Mængde 1 g
Formel vægt 315.37
Optisk rotation −45° (c=1 in Ethanol)
Procent renhed 99%