Få mere at vide
N-Boc-(1S)-3-[3-(3-(isopropyl-d6)-5-methyl-4H-1,2,4-triazol-4-yl)-exo-8-azabicyclo[3.2.1]oct-8-yl]-1-phenyl-1-propanamine, TRC
Artikelnummer. 30337444
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30337444 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337444 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6567472.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Boc-(1S)-3-[3-(3-(isopropyl-d6)-5-methyl-4H-1,2,4-triazol-4-yl)-exo-8-azabicyclo[3.2.1]oct-8-yl]-1-phenyl-1-propanamine
CAS 1217525-03-5
Molekylær formel C27H35D6N5O2
Mængde 2.5 mg
Synonym [(1S)-3-[(3-exo)-3-[3-Methyl-5-(1-methylethyl-d6)-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]oct-8-yl]-1-phenylpropyl]carbamic Acid 1,1-Dimethylethyl Ester, tert-Butyl (1S)-3-[3-(3-(Isopropyl-d6)-5-methyl-4H-1,2,4-triazol-4-yl)-exo-8-azabicyclo[3.2.1]oct-8-yl]-1-phenylpropylcarbamate
InChI formel InChI=1S/C27H41N5O2/c1-18(2)25-30-29-19(3)32(25)23-16-21-12-13-22(17-23)31(21)15-14-24(20-10-8-7-9-11-20)28-26(33)34-27(4,5)6/h7-11,18,21-24H,12-17H2,1-6H3,(H,28,33)/t21?,22?,23?,24-/m0/s1/i1D3,2D3
SMIL [2H]C([2H])([2H])C(c1nnc(C)n1C2CC3CCC(C2)N3CC[C@H](NC(=O)OC(C)(C)C)c4ccccc4)C([2H])([2H])[2H]
IUPAC navn tert-butyl N-[(1S)-3-[3-[3-(1,1,1,3,3,3-hexadeuteriopropan-2-yl)-5-methyl-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octan-8-yl]-1-phenylpropyl]carbamate
Molekylvægt (g/mol) 473.68
Formel vægt 473.3637