missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-1-benzyloxymethyl-L-histidine, 97%

Brand:  Alfa Aesar™ H62663.03

Additional Details : CAS Number : 83468-83-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479158

  • 428.83 DKK / 1g


Estimated Shipment
29-10-2020
Add to basket

Specifications

Chemical Identifiers

83468-83-1
375.425
IEAOAWZLUGOPJX-INIZCTEOSA-N
11245813
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)COCC2=CC=CC=C2)C(=O)O
C19H25N3O5
MFCD00038627
boc-l-his bom-oh, n-boc-n'-benzyloxymethyl-l-histidine, s-3-1-benzyloxy methyl-1h-imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, bochis bom oh, pubchem18939, n alpha-tert-butyloxycarbonyl-n pi-benzyloxyhistidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-phenylmethoxy methyl, 2s-3-1-benzyloxymethyl imidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-1-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid

Specifications

N-Boc-1-benzyloxymethyl-L-histidine
C19H25N3O5
boc-l-his bom-oh, n-boc-n'-benzyloxymethyl-l-histidine, s-3-1-benzyloxy methyl-1h-imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, bochis bom oh, pubchem18939, n alpha-tert-butyloxycarbonyl-n pi-benzyloxyhistidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-phenylmethoxy methyl, 2s-3-1-benzyloxymethyl imidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-1-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)COCC2=CC=CC=C2)C(=O)O
375.425
375.42
97%
1g
83468-83-1
MFCD00038627
IEAOAWZLUGOPJX-INIZCTEOSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid
11245813
Powder
White
Videos
SDS Documents