missing translation for 'onlineSavingsMsg'
Learn More

N-Biotinylcaproylaminocaproylaminocaproylaminoethyl Methanethiosulfonate, TRC

Product Code. 30329543
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30329543 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329543 Supplier Toronto Research Chemicals Supplier No. TRCB39571550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Biotinylcaproylaminocaproylaminocaproylaminoethyl Methanethiosulfonate
CAS 1322625-00-2
Molekylær formel C31H56N6O7S3
Mængde 50 mg
Synonym MTSEA-BIOTINCAPCAPCAP
InChI formel InChI=1S/C31H56N6O7S3/c1-47(43,44)46-22-21-35-29(41)16-7-4-12-19-33-27(39)14-5-2-10-18-32-26(38)15-6-3-11-20-34-28(40)17-9-8-13-25-30-24(23-45-25)36-31(42)37-30/h24-25,30H,2-23H2,1H3,(H,32,38)(H,33,39)(H,34,40)(H,35,41)(H2,36,37,42)
SMIL CS(=O)(=O)SCCNC(=O)CCCCCNC(=O)CCCCCNC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
IUPAC navn N-[6-(2-methylsulfonylsulfanylethylamino)-6-oxohexyl]-6-[6-[5-(2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl)pentanoylamino]hexanoylamino]hexanamide
Molekylvægt (g/mol) 721.01
Formel vægt 720.3373
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.