missing translation for 'onlineSavingsMsg'
Learn More

N-Biotinyl p-Aminophenyl Arsenic Acid, TRC

Product Code. 30344595
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30344595 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344595 Supplier Toronto Research Chemicals Supplier No. TRCB39497010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Biotinyl p-Aminophenyl Arsenic Acid
CAS 212391-23-6
Renhedsgrad noter HPLC
Molekylær formel C16H22AsN3O4S
Mængde 10 mg
Synonym As-[4-[[5-[(3aS,4, S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]phenyl]arsenic Acid
SMIL O=C(NC1=CC=C([As](O)O)C=C1)CCCC[C@H]2C3C(NC(N3)=O)CS2
Molekylvægt (g/mol) 427.35
Formel vægt 427.0547
Procent renhed >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.