Få mere at vide
N-Benzyloxycarbonyl (S)-Lisinopril-d5 Ethyl Methyl Diester, TRC
Artikelnummer. 30331487
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30331487 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331487 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2857622.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Benzyloxycarbonyl (S)-Lisinopril-d5 Ethyl Methyl Diester
CAS 1356383-20-4
Molekylær formel C32H38D5N3O7
Mængde 2.5 mg
Synonym (S)-1-[N2-(1-Carboxy-3-phenylpropyl)-N-benzyloxycarbonyl-L-lysyl]-L-proline-d5 Ethyl Methyl Diester
InChI formel InChI=1S/C32H43N3O7/c1-3-41-30(37)27(20-19-24-13-6-4-7-14-24)34-26(29(36)35-22-12-18-28(35)31(38)40-2)17-10-11-21-33-32(39)42-23-25-15-8-5-9-16-25/h4-9,13-16,26-28,34H,3,10-12,17-23H2,1-2H3,(H,33,39)/t26-,27-,28-/m0/s1/i4D,6D,7D,13D,14D
SMIL [2H]c1c([2H])c([2H])c(CC[C@H](N[C@@H](CCCCNC(=O)OCc2ccccc2)C(=O)N3CCC[C@H]3C(=O)OC)C(=O)OCC)c([2H])c1[2H]
IUPAC navn methyl (2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-(2,3,4,5,6-pentadeuteriophenyl)butan-2-yl]amino]-6-(phenylmethoxycarbonylamino)hexanoyl]pyrrolidine-2-carboxylate
Molekylvægt (g/mol) 586.73
Formel vægt 586.3415