missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-glutaminylglycine, 98%, Thermo Scientific Chemicals

Catalog Number p-7056269
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
6610-42-0
C15H19N3O6
337.332
MFCD00055926
SOUXAAOTONMPRY-NSHDSACASA-N
z-gln-gly-oh, s-2-5-amino-2-benzyloxy carbonyl amino-5-oxopentanamido acetic acid, benzyloxycarbonyl-l-glutaminylglycine, cbz-l-gln-gly-oh, benzyloxy carbonyl-l-glutaminylglycine, s-2-5-amino-2-benzyloxy carbonyl-amino-5-oxopentanamido acetic acid, 2s-2-benzyloxy carbonyl amino-4-carbamoylbutanamido acetic acid, 2-2s-5-amino-5-oxo-2-phenylmethoxycarbonylamino pentanoyl amino acetic acid
6994993
2-[[(2S)-5-amino-5-oxo-2-(phenylmethoxycarbonylamino)pentanoyl]amino]acetic acid
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)N)C(=O)NCC(=O)O
This item is not returnable. View return policy
6610-42-0
C15H19N3O6
337.332
MFCD00055926
SOUXAAOTONMPRY-NSHDSACASA-N
z-gln-gly-oh, s-2-5-amino-2-benzyloxy carbonyl amino-5-oxopentanamido acetic acid, benzyloxycarbonyl-l-glutaminylglycine, cbz-l-gln-gly-oh, benzyloxy carbonyl-l-glutaminylglycine, s-2-5-amino-2-benzyloxy carbonyl-amino-5-oxopentanamido acetic acid, 2s-2-benzyloxy carbonyl amino-4-carbamoylbutanamido acetic acid, 2-2s-5-amino-5-oxo-2-phenylmethoxycarbonylamino pentanoyl amino acetic acid
6994993
2-[[(2S)-5-amino-5-oxo-2-(phenylmethoxycarbonylamino)pentanoyl]amino]acetic acid
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)N)C(=O)NCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 2179068
Formula Weight 337.33
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-L-glutaminylglycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'