Få mere at vide
N-Benzyloxycarbonyl-L-aspartic acid 4-benzyl ester, 98+%, Thermo Scientific™
Artikelnummer. 11378165
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11378165 5 g 5g
11368165 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11378165 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L08436.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Denne Thermo Scientific mærkevare var oprindeligt en del af Alfa Aesar produktportefølje. Noget dokumentation og etiketoplysninger kan referere til det gamle mærke. Originalen Alfa Aesar produkt-/varekode eller SKU-reference er ikke ændret som en del af mærkeovergangen til Thermo Scientific .
Applications
It is widely used as a pharmaceutical intermediate and in food industry.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3479-47-8
Molekylær formel C19H19NO6
Molekylvægt (g/mol) 357.36
MDL nummer MFCD00037820
InChI nøgle VUKCNAATVIWRTF-UHFFFAOYNA-N
Synonym z-asp obzl-oh, cbz-asp obzl-oh, n-cbz-l-aspartic acid 4-benzyl ester, z-l-aspartic acid 4-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-benzyl ester, 4-benzyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid, n-carbobenzoxy-l-aspartic acid 4-benzyl ester, 2s-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid, 2s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid
PubChem CID 47602
IUPAC navn (2S)-4-oxo-4-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMIL OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
Tekniske data
Kemisk navn eller materiale N-Benzyloxycarbonyl-L-aspartic acid 4-benzyl ester
Smeltepunkt 105°C to 107°C
Mængde 5 g
Beilstein 2065292
Opløselighedsinformation Insoluble in water.
Formel vægt 357.37
Procent renhed ≥98%