missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-aspartic acid 1-methyl ester, 98%, Thermo Scientific™

Catalog Number p-7040006
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4668-42-2
C13H15NO6
281.264
MFCD00083277
MFFFBNAPQRDRQW-JTQLQIEISA-N
z-asp-ome, cbz-asp-ome, s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-4-methoxy-4-oxo-3-phenylmethoxycarbonylamino butanoic acid, n-cbz-l-aspartic acid 1-methyl ester, n-cbz-l-asp-ome, z-l-asp-ome, pubchem18988
7021783
(3S)-4-methoxy-4-oxo-3-(phenylmethoxycarbonylamino)butanoic acid
COC(=O)C(CC(=O)O)NC(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
4668-42-2
C13H15NO6
281.264
MFCD00083277
MFFFBNAPQRDRQW-JTQLQIEISA-N
z-asp-ome, cbz-asp-ome, s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-3-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 3s-4-methoxy-4-oxo-3-phenylmethoxycarbonylamino butanoic acid, n-cbz-l-aspartic acid 1-methyl ester, n-cbz-l-asp-ome, z-l-asp-ome, pubchem18988
7021783
(3S)-4-methoxy-4-oxo-3-(phenylmethoxycarbonylamino)butanoic acid
COC(=O)C(CC(=O)O)NC(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Linear Formula HO2CCH2CH(NHCO2CH2C6H5)CO2CH3
Quantity 1 g
Beilstein 4813538
Formula Weight 281.26
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 1-methyl ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'