missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Benzyloxycarbonyl-L-alanine, TRC

Artikelnummer. 30343918
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343918 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30343918 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28661510G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-Benzyloxycarbonyl-L-alanine
CAS 1142-20-7
Molekylær formel C11 H13 N O4
Mængde 10 g
Synonym L-Alanine, N-[(phenylmethoxy)carbonyl]-, Alanine, N-carboxy-, N-benzyl ester, L- (8CI), L-Alanine, N-carboxy-, N-benzyl ester (6CI,7CI), N-[(Phenylmethoxy)carbonyl]-L-alanine, (2S)-2-[[(Benzyloxy)carbonyl]amino]propanoic acid, (S)-2-(Benzyloxycarbonylamino)propanoic acid, (S)-2-[(Benzyloxycarbonyl)amino]propionic acid, Benzyloxycarbonyl-L-alanine, Cbz-L-Ala-OH, L-Carbobenzoxyalanine, L-Carbobenzyloxyalanine, L-Cbz alanine, L-N-(Benzyloxycarbonyl)alanine, N-(Benzyloxycarbonyl)-(S)-alanine, N-(Carbobenzoxy)-L-alanine, N-Benzoxycarbonyl-L-alanine, N-Carbobenzyloxy-L-alanine, N-[(Benzyloxy)carbonyl]-L-alanine, NSC 156978, NSC 88469, CBZ-L-alanine
InChI formel InChI=1S/C11H13NO4/c1-8(10(13)14)12-11(15)16-7-9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,12,15)(H,13,14)/t8-/m0/s1
SMIL C[C@H](NC(=O)OCc1ccccc1)C(=O)O
IUPAC navn (2S)-2-(phenylmethoxycarbonylamino)propanoic acid
Molekylvægt (g/mol) 223.23
Formel vægt 223.0845
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.