missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-2-cyclohexylglycine, 95%, Thermo Scientific™

Catalog Number p-7055202
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
69901-75-3
C16H21NO4
291.347
MFCD00191083
CUSYTUPJAYLNFQ-AWEZNQCLSA-N
z-chg-oh, s-2-benzyloxy carbonyl amino-2-cyclohexylacetic acid, s-cbz-cyclohexylglycine, cbz-cyclohexyl-l-glycine, pubchem5639, 2s-benzyloxy carbonyl amino cyclohexyl acetic acid, n-benzyloxycarbonyl-2-cyclohexyl-l-glycine, s-benzyloxycarbonylamino-cyclohexyl-acetic acid, s-2-benzyloxy carbonyl amino-2-cyclohexylaceti, 2s-2-benzyloxycarbonylamino-2-cyclohexyl-acetic acid
16218812
(2S)-2-cyclohexyl-2-(phenylmethoxycarbonylamino)acetic acid
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2=CC=CC=C2
This item is not returnable. View return policy
69901-75-3
C16H21NO4
291.347
MFCD00191083
CUSYTUPJAYLNFQ-AWEZNQCLSA-N
z-chg-oh, s-2-benzyloxy carbonyl amino-2-cyclohexylacetic acid, s-cbz-cyclohexylglycine, cbz-cyclohexyl-l-glycine, pubchem5639, 2s-benzyloxy carbonyl amino cyclohexyl acetic acid, n-benzyloxycarbonyl-2-cyclohexyl-l-glycine, s-benzyloxycarbonylamino-cyclohexyl-acetic acid, s-2-benzyloxy carbonyl amino-2-cyclohexylaceti, 2s-2-benzyloxycarbonylamino-2-cyclohexyl-acetic acid
16218812
(2S)-2-cyclohexyl-2-(phenylmethoxycarbonylamino)acetic acid
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 291.35
Percent Purity 95%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-L-2-cyclohexylglycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'