Learn More
Alfa Aesar™ N-Benzyloxycarbonyl-D-aspartic acid 4-tert-butyl ester hydrate, 98%
Brand:  Alfa Aesar™ H63757.03
Additional Details : CAS Number : 71449-08-6 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15408506
  • 997.25 DKK / 1g
Estimated Shipment date
from Supplier 01-08-2018
Add to basket
Specifications
Chemical Identifiers
N-Benzyloxycarbonyl-D-aspartic acid 4-tert-butyl ester hydrate
141.36
Z-D-Asp(OtBu)-OH.H2O
71449-08-6
MFCD00153327
Specifications
N-Benzyloxycarbonyl-D-aspartic acid 4-tert-butyl ester hydrate
Solid
141.36
Z-D-Asp(OtBu)-OH.H2O
71449-08-6
98%
MFCD00153327
1g