missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Benzyloxycarbonyl-D-alanine, 98+%, Thermo Scientific Chemicals

Artikelnummer. 11358125
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11358125 1 g 1g
11368125 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11358125 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L08382.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 26607-51-2
Molekylær formel C11H13NO4
Molekylvægt (g/mol) 223.23
MDL nummer MFCD00063126
InChI nøgle TYRGLVWXHJRKMT-UHFFFAOYNA-N
Synonym n-cbz-d-alanine, z-d-ala-oh, cbz-d-ala-oh, z-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, n-carbobenzyloxy-d-alanine, benzyloxycarbonyl-d-alanine, cbz-d-alanine
PubChem CID 70832
IUPAC navn (2R)-2-(phenylmethoxycarbonylamino)propanoic acid
SMIL CC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Tekniske data

Kemisk navn eller materiale N-Benzyloxycarbonyl-D-alanine
Smeltepunkt 83°C to 86°C
Mængde 1 g
Beilstein 2056163
Optisk rotation +14.5° (c=2 in Acetic Acid)
Formel vægt 223.23
Procent renhed ≥98%

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.