Få mere at vide
N-Benzyloxy Naratriptan-d3, TRC
Artikelnummer. 30329664
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30329664 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329664 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28655225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Benzyloxy Naratriptan-d3
CAS 1794937-02-2
Molekylær formel C24H28D3N3O2S
Mængde 25 mg
Synonym N-Methyl-3-(1-methyl-4-piperidinyl)-1-(phenylmethyl)-1H-indole-5-ethanesulfonamide-d3
InChI formel InChI=1S/C24H31N3O2S/c1-25-30(28,29)15-12-19-8-9-24-22(16-19)23(21-10-13-26(2)14-11-21)18-27(24)17-20-6-4-3-5-7-20/h3-9,16,18,21,25H,10-15,17H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])N1CCC(CC1)c2cn(Cc3ccccc3)c4ccc(CCS(=O)(=O)NC)cc24
IUPAC navn 2-[1-benzyl-3-[1-(trideuteriomethyl)piperidin-4-yl]indol-5-yl]-N-methylethanesulfonamide
Molekylvægt (g/mol) 428.61
Formel vægt 428.2325