missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylethanolamine, 96%, Thermo Scientific Chemicals

Catalog Number p-66413
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
104-63-2
C9H13NO
151.21
MFCD00002840
XNIOWJUQPMKCIJ-UHFFFAOYSA-N
n-benzylethanolamine, 2-benzylamino ethanol, benzylaminoethanol, benzylethanolamine, ethanol, 2-phenylmethyl amino, 2-benzylamino ethan-1-ol, benzyl ethanolamine, n-benzyle ethanol amine, ethanol, 2-benzylamino, unii-48121ms9jm
4348
2-(benzylamino)ethanol
OCCNCC1=CC=CC=C1
This item is not returnable. View return policy
104-63-2
C9H13NO
151.21
MFCD00002840
XNIOWJUQPMKCIJ-UHFFFAOYSA-N
n-benzylethanolamine, 2-benzylamino ethanol, benzylaminoethanol, benzylethanolamine, ethanol, 2-phenylmethyl amino, 2-benzylamino ethan-1-ol, benzyl ethanolamine, n-benzyle ethanol amine, ethanol, 2-benzylamino, unii-48121ms9jm
4348
2-(benzylamino)ethanol
OCCNCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 153.0°C to 156.0°C (12.0 mmHg)
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2NHCH2CH2OH
Refractive Index 1.5420 to 1.5440
Quantity 100 mL
Beilstein 12, 1040
Specific Gravity 1.06
Solubility Information Solubility in water: miscible with water. Other solubilities: soluble in alcohol
Formula Weight 151.21
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material N-Benzylethanolamine, 96%
Show More Show Less
compliance-icons
Product Identifier
  • N-Benzylethanolamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'