Få mere at vide
N-Benzylacetamide, TRC
Artikelnummer. 30011289
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011289 5 g
30005734 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011289 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2263155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 588-46-5
Molekylær formel C9 H11 N O
Molekylvægt (g/mol) 149.19
Synonym N-(Phenylmethyl)acetamide, N-Benzylacetamide, Lacosamide Imp. G (Pharmeuropa), Acetamide, N-(phenylmethyl)-, Acetamide, N-benzyl- (6CI,7CI,8CI), N-Acetylbenzylamine, NSC 8065
IUPAC navn N-benzylacetamide
SMIL CC(=O)NCc1ccccc1
Tekniske data
Kemisk navn eller materiale N-Benzylacetamide
Mængde 5 g
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C9H11NO/c1-8(11)10-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3,(H,10,11)
Formel vægt 149.0841