Få mere at vide
N-Benzyl N-Demethyl Trimebutine-d5, TRC
Artikelnummer. 30342761
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30342761 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342761 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2345921MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Benzyl N-Demethyl Trimebutine-d5
CAS 1330189-05-3
Molekylær formel C28H28D5NO5
Mængde 1 mg
Synonym 3,4,5-Trimethoxybenzoic Acid 2-(N-Benzyl-methylamino)-2-phenylbutyl-d5 Ester, N-Benzyl N-Monodesmethyltrimebutine-d5
InChI formel InChI=1S/C28H33NO5/c1-6-28(23-15-11-8-12-16-23,29(2)19-21-13-9-7-10-14-21)20-34-27(30)22-17-24(31-3)26(33-5)25(18-22)32-4/h7-18H,6,19-20H2,1-5H3/i1D3,6D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C(COC(=O)c1cc(OC)c(OC)c(OC)c1)(N(C)Cc2ccccc2)c3ccccc3
IUPAC navn [2-[benzyl(methyl)amino]-3,3,4,4,4-pentadeuterio-2-phenylbutyl] 3,4,5-trimethoxybenzoate
Molekylvægt (g/mol) 468.6
Formel vægt 468.2673