Få mere at vide
N-Benzyl-L-serine, Methyl Ester, TRC
Artikelnummer. 30338967
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30338967 500 mg
1 options
Artikelnummer. 30338967 Leverandør Toronto Research Chemicals Leverandørnr. TRCB289305500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Benzyl-L-Serine, Methyl Ester
CAS 123639-56-5
Molekylær formel C11 H15 N O3
Mængde 500 mg
Synonym L-Serine, N-(phenylmethyl)-, methyl ester, (S)-Methyl 2-(benzylamino)-3-hydroxypropanoate, N-Benzyl-L-serine methyl ester, (S)-N-Benzylserine methyl ester, Methyl N-benzyl-L-serinate, Methyl (2S)-2-(benzylamino)-3-hydroxypropanoate
InChI formel InChI=1S/C11H15NO3/c1-15-11(14)10(8-13)12-7-9-5-3-2-4-6-9/h2-6,10,12-13H,7-8H2,1H3/t10-/m0/s1
SMIL COC(=O)[C@H](CO)NCc1ccccc1
IUPAC navn methyl (2S)-2-(benzylamino)-3-hydroxypropanoate
Molekylvægt (g/mol) 209.24
Formel vægt 209.1052