Learn More
Alfa Aesar™ N-Benzyl-3,5-difluorobenzamide, 97%
Brand:  Alfa Aesar™ H61630.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15408188
  • 4807.66 DKK / 1g
Estimated Shipment date
from Supplier 31-01-2018
Add to basket
Specifications
Chemical Identifiers
N-Benzyl-3,5-difluorobenzamide
247.24
MFCD04806202
Specifications
N-Benzyl-3,5-difluorobenzamide
97%
MFCD04806202
Solid
247.24
1g