missing translation for 'onlineSavingsMsg'
Learn More

N-Benzy N,N-Didesmethyl Trimebutine, TRC

Product Code. 30338007
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30338007 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338007 Supplier Toronto Research Chemicals Supplier No. TRCB27569010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Benzy N,N-Didesmethyl Trimebutine
CAS 1246817-32-2
Molekylær formel C27H31NO5
Mængde 10 mg
Synonym 3,4,5-Trimethoxybenzoic Acid 2-Phenyl-2-[(phenylmethyl)amino]butyl Ester
InChI formel InChI=1S/C27H31NO5/c1-5-27(22-14-10-7-11-15-22,28-18-20-12-8-6-9-13-20)19-33-26(29)21-16-23(30-2)25(32-4)24(17-21)31-3/h6-17,28H,5,18-19H2,1-4H3
SMIL CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(NCc2ccccc2)c3ccccc3
IUPAC navn [2-(benzylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate
Molekylvægt (g/mol) 449.54
Formel vægt 449.2202
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.